Assay Details
| Gene Symbol: RGD1311805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009638.1 | NP_001009638.1 | 19 - 20 | 1969 | 81 | |
| GenBank mRNA | BC087672.1 | - | 19 - 20 | 1969 | 7317049 | 81 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P16725 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167309 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P16725 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P16725 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P16725 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P16725 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19303 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 65856 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 25586 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10800229 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800232 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800219 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800227 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800233 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800226 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800218 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800221 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800231 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800230 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800236 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800235 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800220 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800234 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800222 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800224 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800223 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800228 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800225 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800217 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10800216 | - | - |
| Rat Genome 230 2.0 Array | 1374047_at | - | - |
| Rat Genome 230 2.0 Array | 1374047_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374047_at | - | - |
| Affymetrix HT Rat Focus Array | 1374047_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1800000392.rn.1 | TC1800000041.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1800000391.rn.1 | TC1800000041.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374047_at | - | - |
| Rat Expression Set 230 | 1374047_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10800216 | - | - |
| Rat Genome U34 Set | rc_AI180249_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1354252 | - | - |
| RatRef-12 Expression BeadChip | ILMN_69539 | - | - |

RGD1311805
Genomic Map