Assay Details
| Gene Symbol: RGD1311805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009638.1 | NP_001009638.1 | 2 - 3 | 259 | 129 | |
| GenBank mRNA | BC087672.1 | - | 2 - 3 | 259 | 7317049 | 129 |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1311805
Genomic Map