Assay Details
| Gene Symbol: Krt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199370.1 | NP_955402.1 | 7 - 8 | 1200 | 76 | |
| GenBank mRNA | AY464139.1 | - | 7 - 8 | 1200 | 76 | |
| BC091106.1 | - | 7 - 8 | 1258 | 7381703 | 76 | |
| BC097497.1 | - | 7 - 8 | 1223 | 7462062 | 76 | |
| M63482.1 | - | 7 - 8 | 1264 | 76 | ||
| S76054.1 | - | 7 - 8 | 1264 | 76 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Krt8
Genomic Map