Assay Details
| Gene Symbol: Ngf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277055.1 | NP_001263984.1 | 2 - 3 | 170 | 114 | |
| XM_006233052.2 | XP_006233114.1 | 2 - 3 | 266 | 114 | ||
| XM_006233053.2 | XP_006233115.1 | 2 - 3 | 201 | 114 | ||
| GenBank mRNA | FQ232560.1 | - | 2 - 3 | 170 | 114 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001277055.1,XM_006233052.2,XM_006233053.2 transcripts.
Set Membership: |
|

Ngf
Genomic Map