Assay Details
| Gene Symbol: Them4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025017.1 | NP_001020188.1 | 1 - 2 | 152 | 57 | |
| GenBank mRNA | BC093385.1 | - | 1 - 2 | 152 | 7389383 | 57 |
More Information
Updated Mapping Information
Set Membership: |
|

Them4
Genomic Map