Assay Details
| Gene Symbol: Nupl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139091.2 | NP_620791.1 | 10 - 11 | 1196 | 91 | |
| XM_006252093.2 | XP_006252155.1 | 10 - 11 | 1301 | 91 | ||
| GenBank mRNA | AF000900.1 | - | 9 - 10 | 1067 | 91 | |
| AF000901.1 | - | 9 - 10 | 1089 | 91 | ||
| BC085690.1 | - | 10 - 11 | 1196 | 7190650 | 91 | |
| U63839.1 | - | 10 - 11 | 1209 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nupl1
Genomic Map