Assay Details
| Gene Symbol: Nupl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139091.2 | NP_620791.1 | 5 - 6 | 726 | 141 | |
| XM_006252093.2 | XP_006252155.1 | 5 - 6 | 831 | 141 | ||
| GenBank mRNA | AF000901.1 | - | 4 - 5 | 619 | 141 | |
| BC085690.1 | - | 5 - 6 | 726 | 7190650 | 141 | |
| U63839.1 | - | 5 - 6 | 739 | 141 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF000901.1 transcripts.
Set Membership: |
|

Nupl1
Genomic Map