Assay Details
| Gene Symbol: Nupl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139091.2 | NP_620791.1 | 8 - 9 | 1001 | 151 | |
| XM_006252093.2 | XP_006252155.1 | 8 - 9 | 1106 | 151 | ||
| GenBank mRNA | AF000900.1 | - | 7 - 8 | 872 | 151 | |
| BC085690.1 | - | 8 - 9 | 1001 | 7190650 | 151 | |
| U63839.1 | - | 8 - 9 | 1014 | 151 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nupl1
Genomic Map