Assay Details
| Gene Symbol: RGD1311747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169105.1 | NP_001162576.1 | 1 - 2 | 87 | 107 | |
| GenBank mRNA | DY557818.1 | - | 1 - 2 | 87 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001169105.1 transcripts.
Set Membership: |
|

RGD1311747
Genomic Map