Assay Details
| Gene Symbol: RGD1311747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169105.1 | NP_001162576.1 | 3 - 4 | 443 | 159 | |
| GenBank mRNA | DY557818.1 | - | 3 - 4 | 443 | 159 | |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1311747
Genomic Map