Assay Details
| Gene Symbol: Mrpl54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106770.1 | NP_001100240.1 | 1 - 2 | 162 | 135 | |
| GenBank mRNA | FQ211839.1 | - | 1 - 2 | 145 | 135 | |
| FQ217217.1 | - | 1 - 2 | 145 | 135 | ||
| FQ221492.1 | - | 1 - 2 | 149 | 135 | ||
| FQ224331.1 | - | 1 - 2 | 149 | 135 | ||
| FQ224393.1 | - | 1 - 2 | 145 | 135 | ||
| FQ224740.1 | - | 1 - 2 | 147 | 135 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mrpl54
Genomic Map