Assay Details
| Gene Symbol: Acss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106524.1 | NP_001099994.1 | 7 - 8 | 1262 | 57 | |
| XM_008762307.1 | XP_008760529.1 | 6 - 7 | 2152 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acss1
Genomic Map