Assay Details
| Gene Symbol: Prss54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013888.1 | NP_001013910.1 | 2 - 3 | 411 | 109 | |
| XM_006255073.2 | XP_006255135.1 | 2 - 3 | 427 | 109 | ||
| XM_008772273.1 | XP_008770495.1 | 2 - 3 | 298 | 109 | ||
| GenBank mRNA | BC079217.1 | - | 2 - 3 | 411 | 7131896 | 109 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC079217.1,NM_001013888.1,XM_006255073.2,XM_008772273.1 transcripts.
Set Membership: |
|

Prss54
Genomic Map