Assay Details
| Gene Symbol: Ccl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057151.1 | NP_476492.1 | 2 - 3 | 101 | ||
| GenBank mRNA | AF312687.1 | - | 2 - 3 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccl17
Genomic Map