Assay Details
| Gene Symbol: Irx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107330.1 | NP_001100800.1 | 2 - 3 | 490 | 60 | |
| XM_006227779.2 | XP_006227841.1 | 3 - 4 | 662 | 60 | ||
| XM_008758686.1 | XP_008756908.1 | 2 - 3 | 580 | 60 | ||
| XM_008758687.1 | XP_008756909.1 | 3 - 4 | 486 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Irx4
Genomic Map