Assay Details
| Gene Symbol: Irx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107330.1 | NP_001100800.1 | 3 - 4 | 744 | 62 | |
| XM_006227779.2 | XP_006227841.1 | 4 - 5 | 916 | 62 | ||
| XM_008758685.1 | XP_008756907.1 | 1 - 2 | 715 | 62 | ||
| XM_008758686.1 | XP_008756908.1 | 3 - 4 | 834 | 62 | ||
| XM_008758687.1 | XP_008756909.1 | 4 - 5 | 740 | 62 | ||
| XM_008758688.1 | XP_008756910.1 | 1 - 2 | 87 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008758688.1 transcripts.
Set Membership: |
|

Irx4
Genomic Map