Assay Details
| Gene Symbol: Irx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107330.1 | NP_001100800.1 | 4 - 5 | 858 | 74 | |
| XM_006227779.2 | XP_006227841.1 | 5 - 6 | 1030 | 74 | ||
| XM_008758685.1 | XP_008756907.1 | 2 - 3 | 829 | 74 | ||
| XM_008758686.1 | XP_008756908.1 | 4 - 5 | 948 | 74 | ||
| XM_008758687.1 | XP_008756909.1 | 5 - 6 | 854 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Irx4
Genomic Map