Assay Details
| Gene Symbol: Hbegf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012945.1 | NP_037077.1 | 4 - 5 | 589 | 85 | |
| GenBank mRNA | L05489.1 | - | 4 - 5 | 589 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hbegf
Genomic Map