Assay Details
| Gene Symbol: Mmp24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031757.1 | NP_113945.1 | 8 - 9 | 1359 | 69 | |
| GenBank mRNA | AB023659.1 | - | 8 - 9 | 1359 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mmp24
Genomic Map