Assay Details
| Gene Symbol: Cyp27a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006245175.1 | XP_006245237.1 | 3 - 4 | 525 | 88 | |
| GenBank mRNA | BC061848.1 | - | 3 - 4 | 689 | 5621918 | 88 |
| M38566.1 | - | 3 - 4 | 690 | 88 | ||
More Information
Set Membership: |
|

Cyp27a1
Genomic Map