Assay Details
| Gene Symbol: Cpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012930.1 | NP_037062.1 | 3 - 4 | 406 | 115 | |
| GenBank mRNA | J05470.1 | - | 3 - 4 | 406 | 115 | |
| U88295.1 | - | 3 - 4 | 344 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cpt2
Genomic Map