Assay Details
| Gene Symbol: Ninj2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021595.2 | NP_067606.1 | 2 - 3 | 402 | 106 | |
| XM_008763255.1 | XP_008761477.1 | 2 - 3 | 912 | 106 | ||
| XM_008763256.1 | XP_008761478.1 | 3 - 4 | 829 | 106 | ||
| XM_008763257.1 | XP_008761479.1 | 3 - 4 | 588 | 106 | ||
| XM_008763258.1 | XP_008761480.1 | 3 - 4 | 739 | 106 | ||
| XM_008763259.1 | XP_008761481.1 | 2 - 3 | 548 | 106 | ||
| GenBank mRNA | AB040815.1 | - | 2 - 3 | 403 | 106 | |
| AF250322.1 | - | 2 - 3 | 403 | 106 | ||
More Information
Set Membership: |
|

Ninj2
Genomic Map