Assay Details
| Gene Symbol: Manea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080785.2 | NP_542963.2 | 2 - 3 | 657 | 120 | |
| XM_006237960.1 | XP_006238022.1 | 3 - 4 | 961 | 120 | ||
| XM_006237961.2 | XP_006238023.1 | 3 - 4 | 880 | 120 | ||
| GenBank mRNA | AY599499.1 | - | 2 - 3 | 657 | 120 | |
| FQ223109.1 | - | 3 - 4 | 886 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Manea
Genomic Map