Assay Details
Gene Symbol: Pafah1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031763.3 | NP_113951.1 | 1066 | 108 | ||
GenBank mRNA | AF016049.1 | - | 881 | 108 | ||
BC072510.1 | - | 1066 | 7104733 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
