Assay Details
| Gene Symbol: Tor1aip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199100.2 | NP_954531.1 | 3 - 4 | 1088 | 73 | |
| XM_006250043.2 | XP_006250105.1 | 2 - 3 | 993 | 73 | ||
| XM_006250044.2 | XP_006250106.1 | 2 - 3 | 991 | 73 | ||
| XM_006250045.2 | XP_006250107.1 | 2 - 3 | 991 | 73 | ||
| GenBank mRNA | BC061831.1 | - | 3 - 4 | 1045 | 5599615 | 73 |
More Information
Updated Mapping Information
Set Membership: |
|

Tor1aip2
Genomic Map