Assay Details
| Gene Symbol: Fam227b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013920.2 | NP_001013942.2 | 12 - 13 | 1148 | 66 | |
| XM_006234909.2 | XP_006234971.1 | 12 - 13 | 1071 | 66 | ||
| XM_008762153.1 | XP_008760375.1 | 12 - 13 | 1105 | 66 | ||
| GenBank mRNA | BC079426.1 | - | 12 - 13 | 1107 | 7114265 | 66 |
| BC089861.1 | - | 12 - 13 | 1104 | 7114720 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P158384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380697 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625751 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323804 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246148 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285802 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92569 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354729 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521629 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229258 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947639 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470293 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72386 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87701 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P158384 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P158384 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P158384 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 52607 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 74923 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 93033 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 64437 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 72020 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49871 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 70408 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14062 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18087 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85947 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 12636 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91998 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10849484 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10849482 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10849476 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10849477 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10849478 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10849479 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10849485 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10849475 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10849486 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10849483 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10849481 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10849480 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10849474 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10849473 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10849487 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10849472 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300031589.rn.1 | TC0300004082.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300031596.rn.1 | TC0300004082.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300031590.rn.1 | TC0300004082.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300031592.rn.1 | TC0300004082.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300031593.rn.1 | TC0300004082.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300031591.rn.1 | TC0300004082.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10849472 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22067643 | - | - |

Fam227b
Genomic Map