Assay Details
| Gene Symbol: Pramel6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109342.1 | NP_001102812.1 | 2 - 3 | 541 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pramel6| Gene Symbol: Pramel6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109342.1 | NP_001102812.1 | 2 - 3 | 541 | 89 | |
Set Membership: |
|