Assay Details
| Gene Symbol: Gstcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107725.1 | NP_001101195.1 | 4 - 5 | 1311 | 64 | |
| XM_006233324.2 | XP_006233386.1 | 4 - 5 | 1392 | 64 | ||
| XM_008761502.1 | XP_008759724.1 | 3 - 4 | 2145 | 64 | ||
| XM_008761503.1 | XP_008759725.1 | 4 - 5 | 2968 | 64 | ||
| XM_008761504.1 | XP_008759726.1 | 4 - 5 | 2963 | 64 | ||
| XM_008761505.1 | XP_008759727.1 | 4 - 5 | 2926 | 64 | ||
| GenBank mRNA | FQ211741.1 | - | 4 - 5 | 1397 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P728887 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P452546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172785 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344127 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522302 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643109 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9386 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903962 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242652 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89778 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P728887 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P452546 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P728887 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P452546 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P728887 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P452546 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P452546 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 28940 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 74320 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 56830 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63031 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10826881 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826882 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826877 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826876 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826879 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826873 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826872 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826875 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826878 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826874 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826871 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10826870 | - | - |
| Rat Genome 230 2.0 Array | 1382062_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382062_at | - | - |
| Affymetrix HT Rat Focus Array | 1382062_at | - | - |
| Affymetrix HT Rat Focus Array | 1382062_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200032863.rn.1 | TC0200004726.rn.1 | RTA TC |
| Rat Expression Set 230 | 1382062_PM_at | - | - |
| Rat Expression Set 230 | 1382062_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826870 | - | - |
| Rat Genome U34 Set | rc_AA943385_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21950203 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367890 | - | - |

Gstcd
Genomic Map