Assay Details
| Gene Symbol: Klrb1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085403.1 | NP_001078872.1 | 4 - 5 | 418 | 150 | |
| XM_008763329.1 | XP_008761551.1 | 5 - 6 | 528 | 150 | ||
| XM_008763330.1 | XP_008761552.1 | 5 - 6 | 801 | 150 | ||
| XM_008763331.1 | XP_008761553.1 | 3 - 4 | 820 | 150 | ||
| GenBank mRNA | EF100679.1 | - | 4 - 5 | 418 | 150 | |
| EF100685.1 | - | 4 - 5 | 418 | 150 | ||
| X97477.1 | - | 4 - 5 | 418 | 150 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Klrb1c
Genomic Map