Assay Details
| Gene Symbol: Vwa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013938.1 | NP_001013960.1 | 1 - 2 | 117 | 109 | |
| GenBank mRNA | BC081983.1 | - | 1 - 2 | 117 | 7128587 | 109 |
More Information
Updated Mapping Information
Set Membership: |
|

Vwa1
Genomic Map