Assay Details
| Gene Symbol: Sh3pxd2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006246109.2 | XP_006246171.1 | 8 - 9 | 729 | 69 | |
| XM_008767621.1 | XP_008765843.1 | 7 - 8 | 1300 | 69 | ||
| XM_008767622.1 | XP_008765844.1 | 7 - 8 | 1300 | 69 | ||
More Information
Set Membership: |
|

Sh3pxd2b
Genomic Map