Assay Details
| Gene Symbol: Rab5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105840.2 | NP_001099310.2 | 3 - 4 | 514 | 81 | |
| GenBank mRNA | BC158856.1 | - | 3 - 4 | 496 | 5599461 | 81 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rab5c
Genomic Map