Assay Details
| Gene Symbol: Myh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135158.1 | NP_001128630.1 | 28 - 29 | 3968 | 114 | |
| XM_008767787.1 | XP_008766009.1 | 28 - 29 | 3978 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Myh1
Genomic Map