Assay Details
| Gene Symbol: Myh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135158.1 | NP_001128630.1 | 32 - 33 | 4630 | 131 | |
| XM_008767787.1 | XP_008766009.1 | 32 - 33 | 4640 | 131 | ||
| GenBank mRNA | S68736.1 | - | 8 - 9 | 1385 | 131 | |
More Information
Updated Mapping Information
Set Membership: |
|

Myh1
Genomic Map