Assay Details
| Gene Symbol: Bzrap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768165.1 | XP_008766387.1 | 1 - 2 | 1271 | 60 | |
| XM_213427.7 | XP_213427.5 | 1 - 2 | 1271 | 60 | ||
| GenBank mRNA | AF199337.1 | - | 1 - 2 | 159 | 60 | |
| AF199338.1 | - | 1 - 2 | 159 | 60 | ||
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P006464 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P080612 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P15799 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P006464 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P15799 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P006464 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P15799 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P15799 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P006464 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P165777 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 67493 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 32291 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 75524 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10737301 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737272 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737308 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737276 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737286 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737298 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737302 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737293 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737303 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737288 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737290 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737309 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737277 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737299 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737279 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737274 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737289 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737297 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737295 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737273 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737306 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737284 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737281 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737294 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737283 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737304 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737291 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737300 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737307 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737278 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737280 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737305 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737275 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737285 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737292 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737287 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737282 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737271 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10737266 | - | - |
| Rat Genome 230 2.0 Array | 1380450_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1380450_at | - | - |
| Rat Expression Set 230 | 1380450_PM_at | - | - |
| Rat Expression Set 230 | 1380450_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10737266 | - | - |
| Rat Genome U34 Set | rc_AI044632_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20875118 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349411 | - | - |
| RatRef-12 Expression BeadChip | ILMN_66341 | - | - |

Bzrap1
Genomic Map