Assay Details
| Gene Symbol: Bzrap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768165.1 | XP_008766387.1 | 20 - 21 | 4378 | 84 | |
| XM_213427.7 | XP_213427.5 | 20 - 21 | 4378 | 84 | ||
| GenBank mRNA | AF199337.1 | - | 20 - 21 | 3272 | 84 | |
| AF199338.1 | - | 21 - 22 | 3095 | 84 | ||
More Information
Set Membership: |
|

Bzrap1
Genomic Map