Assay Details
| Gene Symbol: Tmc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126301.1 | NP_001119773.1 | 1 - 2 | 99 | 67 | |
| GenBank mRNA | BC160873.1 | - | 1 - 2 | 99 | 7366375 | 67 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC160873.1, NM_001126301.1 transcripts.
Set Membership: |
|

Tmc8
Genomic Map