Assay Details
| Gene Symbol: Tmc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126301.1 | NP_001119773.1 | 6 - 7 | 70 | ||
| XM_006247818.2 | XP_006247880.1 | 6 - 7 | 1313 | 70 | ||
| XM_006247819.2 | XP_006247881.1 | 6 - 7 | 1310 | 70 | ||
| XM_006247821.2 | XP_006247883.1 | 6 - 7 | 847 | 70 | ||
| XM_008768392.1 | XP_008766614.1 | 6 - 7 | 838 | 70 | ||
| GenBank mRNA | BC160873.1 | - | 6 - 7 | 7366375 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmc8
Genomic Map