Assay Details
| Gene Symbol: MGC95210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247676.2 | XP_006247738.1 | 4 - 5 | 1888 | 67 | |
| XM_008768330.1 | XP_008766552.1 | 4 - 5 | 2969 | 67 | ||
| GenBank mRNA | BC082057.1 | - | 4 - 5 | 1618 | 7132329 | 67 |
| BC088445.1 | - | 4 - 5 | 1877 | 7126436 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

MGC95210
Genomic Map