Assay Details
| Gene Symbol: Osm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006961.1 | NP_001006962.1 | 2 - 3 | 165 | 81 | |
| GenBank mRNA | AB167521.1 | - | 2 - 3 | 165 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Osm
Genomic Map