Assay Details
| Gene Symbol: Vsig4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025004.1 | NP_001020175.1 | 3 - 4 | 517 | 67 | |
| GenBank mRNA | BC091234.1 | - | 3 - 4 | 517 | 7322659 | 67 |
More Information
Updated Mapping Information
Set Membership: |
|

Vsig4
Genomic Map