Assay Details
| Gene Symbol: Acot9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013960.1 | NP_001013982.1 | 14 - 15 | 1310 | 111 | |
| GenBank mRNA | BC085822.1 | - | 14 - 15 | 1310 | 7126662 | 111 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1013822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212073 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931798 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26340 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52980 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1013822 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1013822 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1013822 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1013822 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 51550 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 29156 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 97400 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 44996 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 12854 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49593 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10938087 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10938085 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10938082 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10938089 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10938080 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10938084 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10938090 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10938079 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10938086 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10938083 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10938081 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10938078 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10938088 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10938077 | - | - |
| Rat Genome 230 2.0 Array | 1379262_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1379262_at | - | - |
| Affymetrix HT Rat Focus Array | 1379262_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1379262_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0X00011180.rn.1 | TC0X00001734.rn.1 | RTA TC |
| Rat Expression Set 230 | 1379262_at | - | - |
| Rat Expression Set 230 | 1379262_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10938077 | - | - |
| Rat Genome U34 Set | rc_AA955482_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22076871 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370027 | - | - |

Acot9
Genomic Map