Assay Details
| Gene Symbol: Rnf219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108389.1 | NP_001101859.1 | 5 - 6 | 700 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P146071 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P487832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618897 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410806 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126812 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39355 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P146071 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P487832 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P487832 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P146071 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P487832 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P146071 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P522879 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49645 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10785740 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10785742 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10785741 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10785738 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10785739 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10785737 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10785736 | - | - |
| Rat Genome 230 2.0 Array | 1380760_at | - | - |
| Rat Genome 230 2.0 Array | 1380760_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1500015585.rn.1 | TC1500002304.rn.1 | RTA TC |
| Rat Expression Set 230 | 1380760_PM_at | - | - |
| Rat Expression Set 230 | 1380760_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10785736 | - | - |
| Rat Genome U34 Set | rc_AI235137_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21572902 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364393 | - | - |

Rnf219
Genomic Map