Assay Details
| Gene Symbol: Acss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107793.1 | NP_001101263.1 | 14 - 15 | 1723 | 64 | |
| XM_006235321.2 | XP_006235383.1 | 15 - 16 | 1830 | 64 | ||
| GenBank mRNA | BC161963.1 | - | 14 - 15 | 1703 | 9028172 | 64 |
| FQ217492.1 | - | 15 - 16 | 1674 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P130683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967651 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93295 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807983 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312318 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627179 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486280 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103408 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P130683 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P130683 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P130683 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P501593 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36989 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19562 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38498 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10841356 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841365 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841349 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841357 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841354 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841366 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841361 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841350 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841359 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841364 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841358 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841360 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841362 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841363 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841352 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841355 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841353 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841351 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10841348 | - | - |
| Rat Genome 230 2.0 Array | 1375944_at | - | - |
| Rat Genome 230 2.0 Array | 1375944_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1375944_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1375944_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300015510.rn.1 | TC0300002119.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300015509.rn.1 | TC0300002119.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300015525.rn.1 | TC0300002119.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300015514.rn.1 | TC0300002119.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300015521.rn.1 | TC0300002119.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300015518.rn.1 | TC0300002119.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300015522.rn.1 | TC0300002119.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300015517.rn.1 | TC0300002119.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300015516.rn.1 | TC0300002119.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300015512.rn.1 | TC0300002119.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300015526.rn.1 | TC0300002119.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300015505.rn.1 | TC0300002119.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300015520.rn.1 | TC0300002119.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300015527.rn.1 | TC0300002119.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300015511.rn.1 | TC0300002119.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300015528.rn.1 | TC0300002119.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300015508.rn.1 | TC0300002119.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300015523.rn.1 | TC0300002119.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300015507.rn.1 | TC0300002119.rn.1 | RTA TC |
| Rat Expression Set 230 | 1375944_at | - | - |
| Rat Expression Set 230 | 1375944_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841348 | - | - |
| Rat Genome U34 Set | rc_AA849497_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1360580 | - | - |

Acss2
Genomic Map