Assay Details
| Gene Symbol: Cables2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006235831.2 | XP_006235893.2 | 4 - 5 | 606 | 73 | |
| XM_008762585.1 | XP_008760807.1 | 4 - 5 | 606 | 73 | ||
| XM_008762586.1 | XP_008760808.1 | 4 - 5 | 606 | 73 | ||
| XM_008762588.1 | XP_008760810.1 | 3 - 4 | 534 | 73 | ||
| XM_008762589.1 | XP_008760811.1 | 4 - 5 | 606 | 73 | ||
| XM_008762590.1 | XP_008760812.1 | 4 - 5 | 606 | 73 | ||
| XM_008762591.1 | XP_008760813.1 | 4 - 5 | 606 | 73 | ||
More Information
Set Membership: |
|

Cables2
Genomic Map