Assay Details
| Gene Symbol: Olfml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013192.1 | NP_001013210.1 | 1 - 2 | 440 | 61 | |
| GenBank mRNA | BC081972.1 | - | 1 - 2 | 440 | 7125978 | 61 |
More Information
Updated Mapping Information
Set Membership: |
|

Olfml1
Genomic Map