Assay Details
| Gene Symbol: Grm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105712.1 | NP_001099182.1 | 2 - 3 | 694 | 71 | |
| XM_008762681.1 | XP_008760903.1 | 1 - 2 | 2387 | 71 | ||
| XM_008762682.1 | XP_008760904.1 | 2 - 3 | 1170 | 71 | ||
| XM_008762683.1 | XP_008760905.1 | 1 - 2 | 2387 | 71 | ||
| GenBank mRNA | M92076.1 | - | 1 - 2 | 612 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Grm3
Genomic Map