Assay Details
| Gene Symbol: Ccdc132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173511.1 | NP_001166982.1 | 5 - 6 | 434 | 96 | |
| XM_006236037.2 | XP_006236099.1 | 5 - 6 | 471 | 96 | ||
| XM_008762704.1 | XP_008760926.1 | 5 - 6 | 471 | 96 | ||
| XM_008762705.1 | XP_008760927.1 | 5 - 6 | 471 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc132
Genomic Map