Assay Details
| Gene Symbol: Fbxo5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106206.1 | NP_001099676.1 | 1 - 2 | 204 | 82 | |
| GenBank mRNA | BC158745.1 | - | 1 - 2 | 144 | 9100726 | 82 |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo5
Genomic Map