Assay Details
| Gene Symbol: Tcf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213561.3 | NP_998726.2 | 1142 | 93 | ||
| GenBank mRNA | BC093374.1 | - | 1069 | 7388599 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Tcf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213561.3 | NP_998726.2 | 1142 | 93 | ||
| GenBank mRNA | BC093374.1 | - | 1069 | 7388599 | 93 | |
Set Membership: |
|